Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726605

Sigma-Aldrich

(R)-(+)-α-Ethylbenzylamine

ChiPros®, produced by BASF, ≥99.0%

Synonym(s):

(R)-(+)-1-Phenylpropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
Beilstein/REAXYS Number:
3029904
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

assay

≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥98.0%

density

0.940 g/mL at 20 °C (lit.)

SMILES string

CC[C@@H](N)c1ccccc1

InChI

1S/C9H13N/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9H,2,10H2,1H3/t9-/m1/s1

InChI key

AQFLVLHRZFLDDV-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

170.6 °F

flash_point_c

77 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service