Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726591

Sigma-Aldrich

(S)-(−)-α,4-Dimethylbenzylamine

ChiPros®, produced by BASF, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)NH2
CAS Number:
Molecular Weight:
135.21
Beilstein/REAXYS Number:
2411644
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Product 726591 is not currently sold in your country. Contact Technical Service

grade

produced by BASF

assay

99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.521 (lit.)

bp

205 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

C[C@H](N)c1ccc(C)cc1

InChI

1S/C9H13N/c1-7-3-5-9(6-4-7)8(2)10/h3-6,8H,10H2,1-2H3/t8-/m0/s1

InChI key

UZDDXUMOXKDXNE-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service