Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726516

Sigma-Aldrich

(1R,2R)-trans-2-Benzyloxycyclohexylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(1R)-trans-2-(Phenylmethoxy)cyclohexanamine, (1R,2R)-1-Amino-2-benzyloxycyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19NO
CAS Number:
Molecular Weight:
205.30
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

produced by BASF

assay

≥98.5% (GC)
99%

optical purity

enantiomeric excess: ≥98.5%

SMILES string

N[C@@H]1CCCC[C@H]1OCc2ccccc2

InChI

1S/C13H19NO/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-3,6-7,12-13H,4-5,8-10,14H2/t12-,13-/m1/s1

InChI key

NTHNRYLIXJZHRZ-CHWSQXEVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service