Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726311

Sigma-Aldrich

(1R,2R)-4-Cyclohexene-1,2-diamine

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2
CAS Number:
Molecular Weight:
112.17
Beilstein/REAXYS Number:
7803057
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

≥97.0% (GC)
≥97.0%

form

solid

optical purity

enantiomeric excess: ≥96.0%

storage temp.

−20°C

SMILES string

N[C@@H]1CC=CC[C@H]1N

InChI

1S/C6H12N2/c7-5-3-1-2-4-6(5)8/h1-2,5-6H,3-4,7-8H2/t5-,6-/m1/s1

InChI key

ZDENNVQHCXLZDN-PHDIDXHHSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service