725501
Bis(methyl-η5−cyclopentadienyl)dimethylhafnium
packaged for use in deposition systems
Synonym(s):
HFCMME, HfD-CO2
About This Item
Recommended Products
form
waxy solid
reaction suitability
core: hafnium
color
white
bp
80-120 °C/0.3-0.5 mmHg (lit.)
mp
60 °C (lit.)
density
1.60 g/mL±0.01 g/mL at 25 °C
SMILES string
C[Hf]C.C[C]1[CH][CH][CH][CH]1.C[C]2[CH][CH][CH][CH]2
InChI
1S/2C6H7.2CH3.Hf/c2*1-6-4-2-3-5-6;;;/h2*2-5H,1H3;2*1H3;
InChI key
BSTQZSKPQICLPX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Packaging
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Can the empty deposition cylinders be refilled?
Yes, we are able to refill these cylinders through our SAFC Hitech group. Please contact our Technical Service department by email at [email protected], and they will forward you to the appropriate SAFC Hitech representative for a quote.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service