Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723797

Sigma-Aldrich

Potassium thiophene-2-trifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3BF3KS
CAS Number:
Molecular Weight:
190.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

mp

245-265 °C

SMILES string

[K+].F[B-](F)(F)c1cccs1

InChI

1S/C4H3BF3S.K/c6-5(7,8)4-2-1-3-9-4;/h1-3H;/q-1;+1

InChI key

LXOCUYMZBWNQQS-UHFFFAOYSA-N

Application

Organotrifluoroborate involved in:
  • A variety of cross-coupling reactions, Negishi cross coupling, Nickel-catalyzed cross-coupling, and Suzuki cross-coupling
  • C-O activation of phenol derivatives

Organotrifluoroborates as versatile and stable boronic acid surrogates.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service