Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723452

Sigma-Aldrich

2-Chlorothiazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2ClNS
CAS Number:
Molecular Weight:
119.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.551

density

1.393 g/mL at 25 °C

SMILES string

Clc1nccs1

InChI

1S/C3H2ClNS/c4-3-5-1-2-6-3/h1-2H

InChI key

KLEYVGWAORGTIT-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

129.9 °F - closed cup

flash_point_c

54.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhiraj Patil et al.
Journal of enzyme inhibition and medicinal chemistry, 30(2), 229-239 (2014-06-19)
Asthma is multifaceted disease where many targets contribute towards its development and progression. Among these, adenosine receptor subtypes play a major role. MCD-KV-10, a novel thiazolo-thiophene was designed and evaluated pre-clinically for its implication in management of asthma. This compound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service