Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723231

Sigma-Aldrich

Octadecyl 2-bromoisobutyrate

95%

Synonym(s):

Stearyl initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43BrO2
CAS Number:
Molecular Weight:
419.48
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

95%

form

liquid

refractive index

n20/D 1.460

density

0.998 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)C(C)(C)Br

InChI

1S/C22H43BrO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25-21(24)22(2,3)23/h4-20H2,1-3H3

InChI key

RBOQJHMBYJZWJL-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coessens, V.; Pintauer, T.; Matyjaszewski, K.
Progress in Polymer Science, 26, 337-337 (2001)
Atom transfer radical polymerization.
K Matyjaszewski et al.
Chemical reviews, 101(9), 2921-2990 (2001-12-26)
"Green" atom transfer radical polymerization: from process design to preparation of well-defined environmentally friendly polymeric materials.
Nicolay V Tsarevsky et al.
Chemical reviews, 107(6), 2270-2299 (2007-05-29)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service