Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720763

Sigma-Aldrich

Ethyl 5-oxazolecarboxylate

96%

Synonym(s):

5-Oxazolecarboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3
CAS Number:
Molecular Weight:
141.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

refractive index

n20/D 1.468

density

1.163 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)c1cnco1

InChI

1S/C6H7NO3/c1-2-9-6(8)5-3-7-4-10-5/h3-4H,2H2,1H3

InChI key

KRMORCCAHXFIHF-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.9 °F - closed cup

flash_point_c

79.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service