Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720690

Sigma-Aldrich

N-Boc-2,3,4,5-tetrahydroazepine

97%

Synonym(s):

1-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydroazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO2
CAS Number:
Molecular Weight:
197.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.476

density

1.027 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CCCCC=C1

InChI

1S/C11H19NO2/c1-11(2,3)14-10(13)12-8-6-4-5-7-9-12/h6,8H,4-5,7,9H2,1-3H3

InChI key

JIGRKDDXGVKCEK-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

210.0 °F - closed cup

flash_point_c

98.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service