Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720380

Sigma-Aldrich

(4S,5S)-2-Chloro-3,4-dimethyl-2,5-diphenyl-1-oxa-3-aza-2-silacyclopentane

97%

Synonym(s):

(1S,2S)-pseudoephedrine phenylchlorosilane, (4S,5S)-2-Chloro-3,4-dimethyl-2,5-diphenyl-1,3,2-oxazasilolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClNOSi
CAS Number:
Molecular Weight:
303.86
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

form

oil

density

1.146 g/mL at 25 °C

SMILES string

C[C@H]1[C@@H](O[Si](Cl)(N1C)c2ccccc2)c3ccccc3

InChI

1S/C16H18ClNOSi/c1-13-16(14-9-5-3-6-10-14)19-20(17,18(13)2)15-11-7-4-8-12-15/h3-13,16H,1-2H3/t13-,16+,20?/m0/s1

InChI key

NSZWPVQENFHZEA-DAHSEZPTSA-N

Application

Leighton′s Strained Silacycles: Powerful Enantioselective Allylation Reagents

  • Friedel-Crafts Alkylation with Acylhydrazones
  • Acylhydrazone-Alkene [3+2] Cycloaddition Reactions

Legal Information

Patent pending. U.S. Patent Application No. 11/810,920

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service