Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

719226

Sigma-Aldrich

3,4-Dibromo-2(5H)-furanone

97%

Synonym(s):

3,4-Dibromofuran-2(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Br2O2
CAS Number:
Molecular Weight:
241.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

90-94 °C

functional group

bromo
ester

SMILES string

BrC1=C(Br)C(=O)OC1

InChI

1S/C4H2Br2O2/c5-2-1-8-4(7)3(2)6/h1H2

InChI key

UBAYSWXSWXORAN-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulisses A Pereira et al.
Chemistry & biodiversity, 12(7), 987-1006 (2015-07-15)
Natural phytotoxins and their synthetic analogs are a potential source of new bioactive compounds for agriculture. Analogs of rubrolides, a class of γ-alkylidene-γ-lactones isolated from different ascidians, have been shown to interfere with the photosynthetic electron-transport chain, yet their activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service