Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718815

Sigma-Aldrich

6-Ethoxy-3-pyridinylboronic acid

Synonym(s):

2-Ethoxy-5-pyridineboronic acid, 6-Ethoxypyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10BNO3
CAS Number:
Molecular Weight:
166.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

116-122 °C

storage temp.

2-8°C

SMILES string

CCOc1ccc(cn1)B(O)O

InChI

1S/C7H10BNO3/c1-2-12-7-4-3-6(5-9-7)8(10)11/h3-5,10-11H,2H2,1H3

InChI key

UONCERAQKBPLML-UHFFFAOYSA-N

Application

Reactant for:
  • Domino rhodium-catalyzed alkyne arylation/palladium-catalyzed N arylation
  • Identification of 2-(4-pyridyl)thienopyridinones as GSK-3β inhibitors
  • Suzuki coupling
  • Preparation of heterobiaryls by Suzuki-Miyaura cross-coupling reaction with aryl or heteroaryl halides catalyzed by a palladium precatalyst and monophosphine ligand
  • Suzuki cross-coupling reactions to yield heteroarylpyridines

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service