Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718378

Sigma-Aldrich

S-[4-[2-[4-(2-Phenylethynyl)phenyl]ethynyl]phenyl] thioacetate

95%

Synonym(s):

Thioacetic acid S-[4-[4-(phenylethynyl)phenyl]ethynyl]benzene-thiol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16OS
CAS Number:
Molecular Weight:
352.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

solid

mp

191-196 °C

SMILES string

CC(=O)Sc1ccc(cc1)C#Cc2ccc(cc2)C#Cc3ccccc3

InChI

1S/C24H16OS/c1-19(25)26-24-17-15-23(16-18-24)14-13-22-11-9-21(10-12-22)8-7-20-5-3-2-4-6-20/h2-6,9-12,15-18H,1H3

InChI key

ZWLJTQTZLWBSFY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Hydrolyzing agents deprotect protected thiol derivatives, obtaining unstable free thiols during complex compound synthesis for self-assembled monolayers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service