Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713902

Sigma-Aldrich

(1R,2R)-trans-1,2-Dihydro-1,2-naphthalenediol

≥96.0% (HPLC)

Synonym(s):

(1R-trans)-1,2-Dihydro-1,2-dihydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein/REAXYS Number:
1366285
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

≥96.0% (HPLC)

optical purity

enantiomeric excess: ≥97.0%

storage temp.

2-8°C

SMILES string

O[C@@H]1C=Cc2ccccc2[C@H]1O

InChI

1S/C10H10O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,9-12H/t9-,10-/m1/s1

InChI key

QPUHWUSUBHNZCG-NXEZZACHSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela M Pampanin et al.
Chemosphere, 144, 1372-1376 (2015-10-23)
Fixed wavelength fluorescence (FF) and synchronous fluorescence scanning (SFS) of fish bile are commonly used methods to analyze for exposure to polycyclic aromatic hydrocarbons (PAHs) from petrogenic and pyrogenic sources. A range of conjugated oxidation products from petrogenic PAHs are
R E Parales et al.
Journal of bacteriology, 180(9), 2337-2344 (1998-05-09)
Bacterial three-component dioxygenase systems consist of reductase and ferredoxin components which transfer electrons from NAD(P)H to a terminal oxygenase. In most cases, the oxygenase consists of two different subunits (alpha and beta). To assess the contributions of the alpha and
N A Leneva et al.
Biochemistry. Biokhimiia, 75(5), 562-569 (2010-07-17)
The ability of Pseudomonas fluorescens 26K strain to utilize naphthalene at concentrations up to 600 mg/liter as the sole source of carbon and energy in mineral liquid media was shown. Using HPLC, TLC, and mass-spectrometry, the intermediates of naphthalene transformation
Angela M McIver et al.
Biotechnology progress, 24(3), 593-598 (2008-05-13)
The selective oxidation of aryl substrates to chiral cis-1,2-dihydrodiols is an industrially important reaction for the production of intermediates that can be used to produce fine chemicals, pharmaceuticals, and many other bioactive natural products. More specifically, the oxidation of naphthalene
Gabriela Chilom et al.
Chemosphere, 56(9), 853-860 (2004-07-21)
The formation of bound residues by naphthalene and its metabolite, cis-naphthalene-1,2-dihydrodiol, in a sediment (1% OC), a silty loam soil (2.9% OC) and a peat (26% OC) was examined. The experiments were carried out under both sterile and nonsterile conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service