Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713422

Sigma-Aldrich

Oxalic acid monoethyl ester

Synonym(s):

Monoethyl oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O4
CAS Number:
Molecular Weight:
118.09
Beilstein/REAXYS Number:
1748673
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.424

storage temp.

−20°C

SMILES string

CCOC(=O)C(O)=O

InChI

1S/C4H6O4/c1-2-8-4(7)3(5)6/h2H2,1H3,(H,5,6)

InChI key

JRMAQQQTXDJDNC-UHFFFAOYSA-N

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service