713228
(8α,9S)-10,11-Dihydro-6′-methoxycinchonan-9-amine trihydrochloride
90%
Synonym(s):
(S)-((2S,4S,5R)-5-Ethylquinuclidin-2-yl)(6-methoxyquinolin-4yl)methanamine trihydrochloride, Dihydroquinine-9-epiamine
About This Item
assay
≥89.0% (HPLC)
89.0-111.0% (AT)
90%
optical activity
[α]/D +65.0±5.0°, c = 1 in 0.1 M HCl
SMILES string
Cl.Cl.Cl.CC[C@H]1CN2CC[C@H]1C[C@H]2[C@@H](N)c3ccnc4ccc(OC)cc34
InChI
1S/C20H27N3O.3ClH/c1-3-13-12-23-9-7-14(13)10-19(23)20(21)16-6-8-22-18-5-4-15(24-2)11-17(16)18;;;/h4-6,8,11,13-14,19-20H,3,7,9-10,12,21H2,1-2H3;3*1H/t13-,14-,19-,20-;;;/m0.../s1
InChI key
QINKDORAGBPFFS-HVHHAJEFSA-N
Packaging
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service