Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712965

Sigma-Aldrich

Diethyl (3-cyano-2-methylallyl)phosphonate

mixture of isomers, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16NO3P
CAS Number:
Molecular Weight:
217.20
Beilstein/REAXYS Number:
2363116
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97%
≥97.0% (sum of isomers)

form

liquid

SMILES string

CCOP(=O)(C\C(C)=C\C#N)OCC

InChI

1S/C9H16NO3P/c1-4-12-14(11,13-5-2)8-9(3)6-7-10/h6H,4-5,8H2,1-3H3/b9-6+

InChI key

OPUURRZDCJCVGT-RMKNXTFCSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service