Skip to Content
MilliporeSigma
All Photos(1)

Documents

710520

Sigma-Aldrich

Cyclopentadienylmolybdenum(V) tetrachloride

95%

Synonym(s):

Tetrachloro(η5-2,4-cyclopentadien-1-yl)molybdenum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl4Mo
CAS Number:
Molecular Weight:
302.85
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

95%

reaction suitability

reagent type: catalyst
core: molybdenum

mp

245-249 °C (dec.)

SMILES string

Cl[Mo](Cl)(Cl)Cl.[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.4ClH.Mo/c1-2-4-5-3-1;;;;;/h1-5H;4*1H;/q;;;;;+4/p-4

InChI key

JYLUAHPKLGKVNL-UHFFFAOYSA-J

Application

Catalyst for polymerization of substituted acetylenes

Reactant for reaction with bismuth alkoxides

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service