Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710369

Sigma-Aldrich

Bis(2-furyl)phosphine chloride

85%

Synonym(s):

Di-2-furanylphosphinous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6ClO2P
CAS Number:
Molecular Weight:
200.56
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

85%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.593

density

1.287 g/mL at 25 °C

functional group

phosphine

SMILES string

ClP(c1ccco1)c2ccco2

InChI

1S/C8H6ClO2P/c9-12(7-3-1-5-10-7)8-4-2-6-11-8/h1-6H

InChI key

IYEUYNXDXNWPJI-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Centrostereogenic 1,3-bidentate ferrocenyldiphosphane ligands
  • Ferrocene-based chiral diphosphine
  • Phosphine containing amino acids

Reactant for:
  • α-Arylation of aldehydes
  • Stereoselective [2,3] sigmatropic rearrangement of acyclic allylic phosphinites
  • Hydrophosphination of functionalized alkenes

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P M Ferreira et al.
Scientific reports, 10(1), 18061-18061 (2020-10-24)
Platelet-derived extracellular vesicles (PDEVs) are the most abundant amongst all types of EVs in the circulation. However, the mechanisms leading to PDEVs release, their role in coagulation and phenotypic composition are poorly understood. PDEVs from washed platelets were generated using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service