Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

707317

Sigma-Aldrich

S-Ethyl acetothioacetate

90%

Synonym(s):

3-Oxobutanethioic acid S-ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2S
CAS Number:
Molecular Weight:
146.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

90%

form

liquid

refractive index

n20/D 1.495

density

1.092 g/mL at 25 °C

SMILES string

CCSC(=O)CC(C)=O

InChI

1S/C6H10O2S/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

OMERQNHZTBWLMZ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

177.1 °F

flash_point_c

80.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service