Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

705071

Sigma-Aldrich

(R)-[2-(S-Isobutyl-S-phenylsulfoximinoyl)phenyl]diphenylphosphene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28NOPS
CAS Number:
Molecular Weight:
457.57
MDL number:
UNSPSC Code:
12352000

assay

97%

optical activity

[α]22/D +113°, c = 0.5 in methylene chloride

mp

105-109 °C

SMILES string

CC(C)C[S@](=O)(=Nc1ccccc1P(c2ccccc2)c3ccccc3)c4ccccc4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service