702870
2-Ethoxy-4,4,6-trimethyl-1,3,2-dioxaborinane
97%
About This Item
Recommended Products
assay
97%
form
liquid
refractive index
n20/D 1.414
density
0.921 g/mL at 25 °C
SMILES string
CCOB1OC(C)CC(C)(C)O1
InChI
1S/C8H17BO3/c1-5-10-9-11-7(2)6-8(3,4)12-9/h7H,5-6H2,1-4H3
InChI key
JOHWACCNDCSHCO-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
120.0 °F
flash_point_c
48.9 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service