Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700169

Sigma-Aldrich

4-(Dimethylamino)pyridine, ChemDose tablets

Loading: 0.04 mmol tablet.

Synonym(s):

ChemDose, 4-(Dimethylamino)pyridine impregnated tablets, DMAP, ChemDose impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CN(C)c1ccncc1

InChI

1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3

InChI key

VHYFNPMBLIVWCW-UHFFFAOYSA-N

Legal Information

Covered by International Patent Application No. PCT/DK05/000428 and related patent applications, including European Patent Application No. 05753717.7.
ChemDose is a trademark of Reaxa Ltd.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 1

target_organs

Nervous system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M van Wijk et al.
Analytical and bioanalytical chemistry, 400(5), 1375-1385 (2011-03-30)
A generic LC-MS/MS method was developed for the analysis of potentially genotoxic alkyl halides. A broad selection of alkyl halides were derivatized using 4-dimethylaminopyridine in acetonitrile. The reaction conditions for derivatization, i.e., solvent, reaction time, temperature and concentration of alkyl
Capture and visualization of hydrogen sulfide by a fluorescent probe.
Chunrong Liu et al.
Angewandte Chemie (International ed. in English), 50(44), 10327-10329 (2011-09-08)
Min Suk Shim et al.
Journal of controlled release : official journal of the Controlled Release Society, 133(3), 206-213 (2008-11-11)
To maximize therapeutic effects, targeted delivery of nucleic acids (e.g., DNA and RNA) in their appropriate intracellular targets is highly desirable. In this study, primary amines of a model polymeric nonviral carrier, polyethylenimine (PEI), at two molecular weights (0.8 and
Chenghu Yan et al.
Biomacromolecules, 10(8), 2013-2018 (2009-09-03)
An effective method for grafting L-lactide (LA) from unmodified cellulose by ring-opening polymerization (ROP) in homogeneous mild conditions is presented. By using 4-dimethylaminopyridine (DMAP) as an organic catalyst, cellulose-graft-poly(L-lactide) (cellulose-g-PLLA) copolymers with a molar substitution (MS(PLLA)) of PLLA in a
Hangxiang Wang et al.
Journal of the American Chemical Society, 133(31), 12220-12228 (2011-07-19)
Catalysts hold promise as tools for chemical protein modification. However, the application of catalysts or catalyst-mediated reactions to proteins has only recently begun to be addressed, mainly in in vitro systems. By radically improving the affinity-guided DMAP (4-dimethylaminopyridine) (AGD) catalysts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service