Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699446

Sigma-Aldrich

3-(4-Morpholinomethyl)phenylboronic acid pinacol ester

95%

Synonym(s):

3-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaboran-2-yl)benzyl]morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26BNO3
CAS Number:
Molecular Weight:
303.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

mp

52-56 °C

functional group

ether

SMILES string

CC1(C)OB(OC1(C)C)c2cccc(CN3CCOCC3)c2

InChI

1S/C17H26BNO3/c1-16(2)17(3,4)22-18(21-16)15-7-5-6-14(12-15)13-19-8-10-20-11-9-19/h5-7,12H,8-11,13H2,1-4H3

InChI key

RCGRLLZELIIKDH-UHFFFAOYSA-N

Application

3-(4-Morpholinomethyl)phenylboronic acid pinacol ester can be used as a reactant:
  • In the palladium-catalyzed Suzuki-Miyaura coupling reaction to synthesize biaryl derivatives.[1]
  • To prepare a morpholinylphenyl anilinoquinazoline lapatinib analog.[2]
  • To synthesize diaryl or heteroaryl triazole quinuclidine derivatives as potent α7 nicotinic acetylcholine receptor ligands.[3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-mediated synthesis of an arylboronate library
Spencer J, et al.
ACS Combinatorial Science, 13(1), 24-31 (2011)
Bis (het) aryl-1, 2, 3-triazole quinuclidines as α-7 nicotinic acetylcholine receptor ligands: Synthesis, structure affinity relationships, agonism activity,[18F]-radiolabeling and PET study in rats
Ouach A, et al.
European Journal of Medicinal Chemistry, 179(10), 449-469 (2019)
Gautam Patel et al.
Journal of medicinal chemistry, 56(10), 3820-3832 (2013-04-20)
Human African trypanosomiasis (HAT) is a neglected tropical disease caused by the protozoan parasite Trypanosoma brucei . Because drugs in use against HAT are toxic and require intravenous dosing, new drugs are needed. Initiating lead discovery campaigns by using chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service