Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698725

Sigma-Aldrich

(3aR,6aR)-3,6-Diphenyl-1,3a,4,6a-tetrahydropentalene

97%

Synonym(s):

(3aR,6aR)-1,3a,4,6a-tetrahydro-3,6-diphenylpentalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18
CAS Number:
Molecular Weight:
258.36
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

97%

form

powder

optical activity

[α]22/D −600, c = 1 in chloroform

mp

104-109 °C

storage temp.

2-8°C

SMILES string

C1C=C([C@@H]2CC=C([C@H]12)c3ccccc3)c4ccccc4

InChI

1S/C20H18/c1-3-7-15(8-4-1)17-11-13-20-18(12-14-19(17)20)16-9-5-2-6-10-16/h1-12,19-20H,13-14H2/t19-,20-/m0/s1

InChI key

IEWYLVULSUMUEM-PMACEKPBSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service