Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697923

Sigma-Aldrich

(R)-2-[(Boc-amino)methyl]-3-methylbutyric acid

97%

Synonym(s):

(R)-N-Boc-2-isopropyl-β-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
MDL number:
UNSPSC Code:
12352200

assay

97%

optical activity

[α]22/D −3.0°, c = 0.5 in methanol

mp

121-126 °C

SMILES string

CC(C)[C@H](CNC(=O)OC(C)(C)C)C(O)=O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service