Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697672

Sigma-Aldrich

(3-Furanyl)sulfur pentafluoride

Synonym(s):

Pentafluoro-3-furanylsulfur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F5OS
CAS Number:
Molecular Weight:
194.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.373

bp

101-103 °C/760 mmHg

density

1.571 g/mL at 25 °C

SMILES string

FS(F)(F)(F)(F)c1ccoc1

InChI

1S/C4H3F5OS/c5-11(6,7,8,9)4-1-2-10-3-4/h1-3H

InChI key

MDJSCQSIEMJGMO-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

55.0 °F

flash_point_c

12.8 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service