Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697435

Sigma-Aldrich

2,2,2-Trifluoroethyl vinyl ether

97%

Synonym(s):

(2,2,2-Trifluoroethoxy)ethene, Fluoroxene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5F3O
CAS Number:
Molecular Weight:
126.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.320

density

1.108 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)COC=C

InChI

1S/C4H5F3O/c1-2-8-3-4(5,6)7/h2H,1,3H2

InChI key

DLEGDLSLRSOURQ-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-20.0 °F

flash_point_c

-28.9 °C

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ARE ALL HALOGENS HEPATOTOXIC?
G M CANNON et al.
Anesthesia and analgesia, 43, 400-406 (1964-07-01)
ANAESTHETIC MANAGEMENT FOR CARDIOPULMONARY BYPASS: A REVIEW OF 200 CASES.
J A PIERCE et al.
Canadian Anaesthetists' Society journal, 12, 179-191 (1965-03-01)
APPLICATIONS OF GAS CHROMATOGRAPHY IN ANESTHESIA.
L W FABIAN
Clinical anesthesia, 3, 145-160 (1964-01-01)
SOLUBILITY OF FLUROXENE IN BLOOD AND TISSUE HOMOGENATES.
E S MUNSON et al.
Anesthesiology, 25, 638-640 (1964-09-01)
[PRELIMINARY EXPERIENCES WITH FLUOROMAR].
O ZAFFIRI et al.
Minerva anestesiologica, 30, 184-187 (1964-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service