697427
2-(Diisopropylphosphino)ethylamine
kanata purity
Synonym(s):
2-[Bis(1-methylethyl)phosphino]ethanamine
About This Item
reaction suitability
reagent type: ligand
refractive index
n20/D 1.489
density
0.872 g/mL at 25 °C
functional group
phosphine
SMILES string
CC(C)P(CCN)C(C)C
InChI
1S/C8H20NP/c1-7(2)10(6-5-9)8(3)4/h7-8H,5-6,9H2,1-4H3
InChI key
QUVUVMRUSAKOPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service