Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696412

Sigma-Aldrich

Ethyl (R)-N-Boc-piperidine-3-carboxylate

95%

Synonym(s):

(R)-(−)-N-Boc-nipecotic acid ethyl ester, (R)-1-(tert-Butoxycarbonyl)-piperidine-3-carboxylic acid ethyl ester, (R)-1-(tert-Butyl) 3-ethyl 1,3-piperidinedicarboxylate, Ethyl (3R)-1-tert-butoxycarbonyl-3-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23NO4
CAS Number:
Molecular Weight:
257.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

36-40 °C

SMILES string

CCOC(=O)[C@@H]1CCCN(C1)C(=O)OC(C)(C)C

InChI

1S/C13H23NO4/c1-5-17-11(15)10-7-6-8-14(9-10)12(16)18-13(2,3)4/h10H,5-9H2,1-4H3/t10-/m1/s1

InChI key

YCXCRFGBFZTUSU-SNVBAGLBSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service