Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695521

Sigma-Aldrich

3-(2-Thienyl)aziridine-2-carboxaldehyde dimer

95%

Synonym(s):

6-(2-Thienyl)-2-[3-(2-thienyl)-2-aziridinyl]-3-oxa-1-azabicyclo[3.1.0]hexan-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2O2S2
CAS Number:
Molecular Weight:
306.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

mp

148-152 °C (D)

SMILES string

O[C@@H]1O[C@@H]([C@H]2N[C@@H]2c3cccs3)N4[C@H]1[C@H]4c5cccs5

InChI

1S/C14H14N2O2S2/c17-14-12-11(8-4-2-6-20-8)16(12)13(18-14)10-9(15-10)7-3-1-5-19-7/h1-6,9-15,17H/t9-,10+,11-,12+,13+,14-,16?/m1/s1

InChI key

RYUJUWWXTHBUSR-JHLSSSLQSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Dam. 1 - Flam. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service