Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695483

Sigma-Aldrich

(3R)-N-Boc-2-azabicyclo[2.2.1]heptane-3-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO4
CAS Number:
Molecular Weight:
241.28
MDL number:
UNSPSC Code:
12352000

assay

97%

optical activity

[α]22/D +163°, c = 1 in chloroform

mp

148-153 °C

SMILES string

CC(C)(C)OC(=O)N1C2CCC(C2)[C@@H]1C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 36, 6765-6765 (1995)
Srikanth Venkatraman et al.
Bioorganic & medicinal chemistry letters, 16(6), 1628-1632 (2006-01-18)
Prolonged hepatitis C infection is the leading cause for cirrhosis of the liver and hepatocellular carcinoma. The etiological agent HCV virus codes a single polyprotein of approximately 3000 amino acids that is processed with the help of a serine protease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service