693766
Cerium
foil, size 25 mm × 25 mm × 1 mm, 99.9% (rare earth, metals basis)
About This Item
assay
99.9% (rare earth, metals basis)
form
foil
resistivity
73 μΩ-cm, 20°C
size
25 mm × 25 mm × 1 mm
bp
3443 °C (lit.)
mp
795 °C (lit.)
density
6.67 g/mL at 25 °C (lit.)
SMILES string
[Ce]
InChI
1S/Ce
InChI key
GWXLDORMOJMVQZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service