Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693561

Sigma-Aldrich

(S)-N-(3,5-Dimethylphenyl)-3-methyl-2-(N-formyl-N-methylamino)butanamide

97%

Synonym(s):

Kenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O2
CAS Number:
Molecular Weight:
262.35
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

optical activity

[α]22/D -229°, c = 1 in chloroform

mp

97-104 °C

storage temp.

2-8°C

SMILES string

CC(C)[C@H](N(C)C=O)C(=O)Nc1cc(C)cc(C)c1

InChI

1S/C15H22N2O2/c1-10(2)14(17(5)9-18)15(19)16-13-7-11(3)6-12(4)8-13/h6-10,14H,1-5H3,(H,16,19)/t14-/m0/s1

InChI key

WWUZCRRIJZUXEZ-AWEZNQCLSA-N

Application

Used in the asymmetric reduction of prochiral N-aryl ketimines with trichlorosilane.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malkov, A. V.; Stoncius, S.; MacDougall, K. N.; Mariani, A.; McGeoch, G. D.; Kocovsky, P.
Tetrahedron, 62, 264-264 (2006)
Enantioselective synthesis of 1,2-diarylaziridines by the organocatalytic reductive amination of alpha-chloroketones.
Andrei V Malkov et al.
Angewandte Chemie (International ed. in English), 46(20), 3722-3724 (2007-04-07)
Andrei V Malkov et al.
Organic letters, 6(13), 2253-2256 (2004-06-18)
[reaction: see text] Asymmetric reduction of ketimines 1 with trichlorosilane can be catalyzed by a new N-methyl L-valine derived Lewis basic organocatalyst, such as 4d, with high enantioselectivity. The structure-reactivity investigation suggests hydrogen bonding and arene-arene interactions between the catalyst
Pengzhen Wang et al.
Annals of translational medicine, 9(11), 940-940 (2021-08-06)
Osteoarthritis (OA) is characterized by erosion and degradation of articular cartilage. This study assessed the effects of curcumin on mouse knee cartilage chondrocytes. Chondrocytes were treated for 24 hours with interleukin IL-1β (10 ng/mL) alone, or the combination of curcumin
Pengzhen Wang et al.
The Knee, 27(1), 18-25 (2019-12-31)
Articular cartilage is a unique avascular tissue in which chondrocytes are embedded in extracellular matrix (ECM). The decreased ECM resulting from the loss of articular chondrocyte viability leads to degenerative diseases such as osteoarthritis (OA). This study aims to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service