Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693324

Sigma-Aldrich

(S)-[(RuCl(H8-BINAP))2(μ-Cl)3][NH2Me2]

Synonym(s):

Dimethylammonium dichlorotri(μ-chloro)bis[(S)-(−)-2,2′-bis(diphenylphosphino)-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl]diruthenate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH2+[C88H80Cl5P4Ru2]-
CAS Number:
Molecular Weight:
1686.97
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

functional group

amine
phosphine

storage temp.

2-8°C

InChI

1S/2C44H40P2.C2H7N.2ClH.3Cl.2Ru/c2*1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;1-3-2;;;;;;;/h2*1-12,19-26,29-32H,13-18,27-28H2;3H,1-2H3;2*1H;;;;;/q;;;;;3*+1;2*-1/p-1

InChI key

BQQWMTMUAYPPMV-UHFFFAOYSA-M

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service