Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

689971

Sigma-Aldrich

N-Phenylbenzimidoyl chloride

95%

Synonym(s):

N-(α-Chlorobenzylidene)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10ClN
CAS Number:
Molecular Weight:
215.68
Beilstein/REAXYS Number:
511861
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥94.5% (GC)
95%

form

lumps

mp

38-43 °C

functional group

chloro
phenyl

storage temp.

−20°C

SMILES string

ClC(=Nc1ccccc1)c2ccccc2

InChI

1S/C13H10ClN/c14-13(11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H

InChI key

WFOFSUPMQDLYOM-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Monier et al.
Carbohydrate polymers, 195, 652-661 (2018-05-29)
A surface molecular imprinting technique was utilized in the fabrication of an enantio-selective adsorbent based on amino-functionalized alginate microspheres for chiral resolution of ascorbic acid. Alginate microspheres were first strengthened via epichlorohydrin (ECH) covalent cross-linking then functionalized with amino groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service