Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

689106

Sigma-Aldrich

Ethyl 4,4,4-trifluoroacetoacetate

Arxada quality, ≥98.0% (GC)

Synonym(s):

Ethyl 3-oxo-4,4,4-trifluorobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCH2CO2C2H5
CAS Number:
Molecular Weight:
184.11
Beilstein/REAXYS Number:
608353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.0% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.20% water (Karl Fischer)

refractive index

n20/D 1.375 (lit.)

bp

129-130 °C (lit.)

density

1.259 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)C(F)(F)F

InChI

1S/C6H7F3O3/c1-2-12-5(11)3-4(10)6(7,8)9/h2-3H2,1H3

InChI key

OCJKUQIPRNZDTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

color values: L*98.00-100.50 (CIELAB)

pictograms

FlameExclamation mark

signalword

Warning

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

100.4 °F - closed cup

flash_point_c

38 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service