Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688908

Sigma-Aldrich

Propionaldehyde oxime

≥96%, ≥96.0% (mixture of isomers)

Synonym(s):

Propanal oxime, Propanaldoxime, Propionaldoxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO
CAS Number:
Molecular Weight:
73.09
Beilstein/REAXYS Number:
1098365
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥96%
≥96.0% (mixture of isomers)

form

liquid

storage temp.

2-8°C

SMILES string

CC\C=N/O

InChI

1S/C3H7NO/c1-2-3-4-5/h3,5H,2H2,1H3/b4-3-

InChI key

IFDZZSXEPSSHNC-ARJAWSKDSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Shezmin Zavahir et al.
The Analyst, 144(16), 4803-4812 (2019-07-06)
This study reports gas chromatography (GC) combined with Fourier transform infrared (FTIR) spectroscopy to investigate the elution profiles of individual oxime isomers undergoing characteristic interconversion (dynamic chromatography) in GC. The use of a light-pipe FTIR interface enables on-line acquisition of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service