Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688118

Sigma-Aldrich

Tris[1-phenylisoquinoline-C2,N]iridium(III)

sublimed grade

Synonym(s):

Ir(piq)3, Tris[1-phenylisoquinolinato-C2,N]iridium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H30IrN3
CAS Number:
Molecular Weight:
804.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

sublimed grade

loss

0.5% loss on heating, 410°C (typical, TGA)

mp

440 °C
454 °C

λmax

324 nm

fluorescence

λex 324 nm; λem 615 nm in THF

OLED device performance

ITO/PEDOT:PSS/NPD/CBP:Ir(piq)3 (6 wt%)/Alq3/LiF/Al

  • Color: red
  • Max. Luminance: 20000 Cd/m2
  • Max. EQE: 8.1 %
  • Turn-On Voltage: 5.9 V

SMILES string

[Ir](C1=CC=CC=C1C2=NC=CC3=C2C=CC=C3)(C4=CC=CC=C4C5=NC=CC6=C5C=CC=C6)C7=CC=CC=C7C8=NC=CC9=C8C=CC=C9

InChI

1S/3C15H10N.Ir/c3*1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15;/h3*1-7,9-11H;

InChI key

DGLAKLWHAUIQCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific traces available upon request

Application

OLED triplett emitter (red)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient and stable organic light-emitting diode using 4,4'-bis(N-carbazolyl)-9,9'-spirobifluorene as a thermally stable host material
Tsuzuki, T.; Tokito, S.
Applied Physics Letters, 94, 033302-033302 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service