Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

685593

Sigma-Aldrich

Palladium(II) acetate, ChemDose tablets

Loading: 10μmol per tablet

Synonym(s):

ChemDose, Palladium(II) acetate tablets, Palladium(II) acetate impregnated tablets, Pd(OAc)2 impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

matrix

Magnesium aluminometasilicate particle platform

SMILES string

CC(=O)O[Pd]OC(C)=O

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChemDose is a trademark of Reaxa Ltd.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How should I dispose of spent ChemDose® tablets?

    They should be treated as solid waste, contaminated with reaction species. The matrix is non-toxic and environmentally benign.

  4. How should I store ChemDose® tablets?

    As with most catalysts and ligands, as a general precaution it is best to store them in a refrigerator and/or under an inert atmosphere to maintain activity. Some catalysts, such as Pd(dppf)Cl2·CH2Cl2, are more robust and can be handled in air without issue. Please refer to the ChemDose® label for specific handling of a given catalyst, ligand, or reagent.

  5. Can I use ChemDose® tablets in any solvent?

    Yes, including aqueous solvents.

  6. Do the ChemDose® tablets disintegrate during the course of the reaction?

    They are normally recovered intact, however, they may disintegrate somewhat in aqueous reactions. Any disintegrated tablets can be removed from the reaction via filtration.

  7. What is the pH stability of the ChemDose® tablets?

    The ChemDose® tablets are mildly basic and will dissolve in the presence of mineral acids. 

  8. Can I use ChemDose® tablets in a microwave or pressure vessel?

    Absolutely. Please see the Technical Information for Download section (below) for additional examples of cross-coupling reactions involving the use of microwaves. 

  9. What is the difference between Product 520764, Palladium (II) acetate, and the palladium acetate in the form of a ChemDose® tablet?

    The ChemDose® products offer a streamlined way to catalyze reactions. The catalyst is dispersed in an inert tablet matrix in specific millimolar and micromolar quantities. The tablets can decrease time spent on weighing/adding the catalyst to the reaction mixture. The tablets effectively give the same reaction yields with a controlled release of the catalyst. They are also extremely easy to remove after the reaction is complete. The inert tablet usually stays intact throughout the reaction. Product 684929 is the ChemDose® tablet that contains 2.0 μmol of palladium (II) acetate, while product#: 685593 is the 10μmol version. More information on these products can be found at the link below. 

  10. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  11. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  12. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Jinyue Ding et al.
Nature communications, 5, 5474-5474 (2014-11-19)
Non-aromatic nitrogen- and oxygen-containing heterocycles such as piperidines and pyrans are prevalent components of natural products and pharmaceutical drugs. Although it has been a workhorse as a synthetic method for assembling unsaturated sp(2)-hybridized substrates, transition metal-catalysed cross-coupling chemistry is traditionally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service