About This Item
Recommended Products
assay
≥97.0% (GC)
refractive index
n20/D 1.402
bp
67-68 °C (lit.)
density
0.692 g/mL at 20 °C (lit.)
SMILES string
CC\C(C)=C/C
InChI
1S/C6H12/c1-4-6(3)5-2/h4H,5H2,1-3H3/b6-4-
InChI key
BEQGRRJLJLVQAQ-XQRVVYSFSA-N
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-16.6 °F - closed cup
flash_point_c
-27 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service