Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682918

Sigma-Aldrich

Dicyclopentylphosphine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19P
CAS Number:
Molecular Weight:
170.23
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

bp

76-78 °C/0.8 mmHg

density

0.933 g/mL at 25 °C

functional group

phosphine

SMILES string

C1CCC(C1)PC2CCCC2

InChI

1S/C10H19P/c1-2-6-9(5-1)11-10-7-3-4-8-10/h9-11H,1-8H2

InChI key

OKMVYXTWPPPSIM-UHFFFAOYSA-N

Application

Reactant for:
  • Interactions with electron deficient boranes
  • Cross-coupling between thiols and secondary phosphines
  • CsOH-promoted P-alkylation for the formation of tertiary phosphines

Reactant for synthesis of:
  • Daniphos ligand
  • Ferrocene-based 1,3-bis(phosphanes) with planar chirality

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Pyr. Liq. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service