Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68290

Sigma-Aldrich

4-Methyl-1,3-pentadiene

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH=CH2
CAS Number:
Molecular Weight:
82.14
Beilstein/REAXYS Number:
1697096
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.0% (GC)

refractive index

n20/D 1.452

bp

75-77 °C

density

0.718 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C\C=C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service