Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681644

Sigma-Aldrich

4-(Trifluoromethyl)-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F3O
CAS Number:
Molecular Weight:
200.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

bp

67-72/0.1 mmHg

mp

38-42 °C

SMILES string

FC(F)(F)c1cccc2C(=O)CCc12

InChI

1S/C10H7F3O/c11-10(12,13)8-3-1-2-7-6(8)4-5-9(7)14/h1-3H,4-5H2

InChI key

LJVBFMQEZSEGRL-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>226.9 °F - closed cup

flash_point_c

> 108.3 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service