Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680834

Sigma-Aldrich

N-(2,2,2-Trichloroethoxysulfonyl)urea

96%

Synonym(s):

Aminocarbonylsulfamic acid, 3,3,3-trichloroethoxy ester, Tces-Urea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONHSO2OCH2CCl3
CAS Number:
Molecular Weight:
271.51
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

solid

mp

161-165 °C

SMILES string

NC(=O)NS(=O)(=O)OCC(Cl)(Cl)Cl

InChI

1S/C3H5Cl3N2O4S/c4-3(5,6)1-12-13(10,11)8-2(7)9/h1H2,(H3,7,8,9)

InChI key

QYUJBOJJEGIBCJ-UHFFFAOYSA-N

Application

  • Reactant for oxidative C-H amination reactions
Reagent for Rh-catalyzed urea formation.[1]
Reagent for cyclic urea synthesis.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihyong Kim et al.
Organic letters, 8(6), 1073-1076 (2006-03-10)
[reaction: see text] Oxidative C-H amination of N-trichloroethoxysulfonyl-protected ureas and guanidines is demonstrated to proceed in high yield for tertiary and benzylic-derived substrates. The success of these reactions is predicated on the choice of the electron-withdrawn 2,2,2-trichloroethoxysulfonyl (Tces) protecting group

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service