Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68080

Sigma-Aldrich

3-Methylnonane

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
142.28
Beilstein/REAXYS Number:
1718850
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99.0% (GC)

refractive index

n20/D 1.413

bp

166-167 °C (lit.)

density

0.732 g/mL at 20 °C (lit.)

SMILES string

CCCCCCC(C)CC

InChI

1S/C10H22/c1-4-6-7-8-9-10(3)5-2/h10H,4-9H2,1-3H3

InChI key

PLZDDPSCZHRBOY-UHFFFAOYSA-N

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service