Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679364

Sigma-Aldrich

Isobutyl(trimethoxy)silane

≥98%, deposition grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2Si(OCH3)3
CAS Number:
Molecular Weight:
178.30
Beilstein/REAXYS Number:
2233976
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

assay

≥98%

form

solution

refractive index

n20/D 1.396 (lit.)

bp

137 °C
154 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CC(C)C)(OC)OC

InChI

1S/C7H18O3Si/c1-7(2)6-11(8-3,9-4)10-5/h7H,6H2,1-5H3

InChI key

XYJRNCYWTVGEEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

80.6 °F - closed cup

flash_point_c

27 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin J Privett et al.
Biofouling, 26(8), 973-983 (2010-11-18)
This report details the efficacy of nitric oxide (NO)-releasing xerogel surfaces composed of N-(6-aminohexyl)aminopropyl trimethoxysilane (AHAP3) and isobutyltrimethoxysilane (BTMOS) against Candida albicans adhesion, viability, and biofilm formation. A parallel plate flow cell assay was used to examine the effect of

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Silica's versatility spans various industries, including biomedical applications.

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service