Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677728

Sigma-Aldrich

Bromo(tri-tert-butylphosphine)palladium(I) dimer

Synonym(s):

Dibromobis(tri-tert-butylphosphine)dipalladium, Dibromobis(tri-tert-butylphosphino)dipalladium(I), Palladium(I) tri-tert-butylphosphine bromide, dimer, Pd-113, [Pd(μ-Br)t-Bu3P]2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Pd(C4H9)3PBr]2
CAS Number:
Molecular Weight:
777.28
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Br[Pd].Br[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

Application

  • Catalyst for γ-arylation of α,β-unsaturated esters from silyl ketene acetals[1] (eq. 1)
  • Catalyst for carbon -sulfur bond formation[2] (eq. 2)
  • Catalyst for diastereoselective arylation of 4-substituted cyclohexyl esters[3] (eq. 3)
  • Catalyst for Suzuki coupling of hindered aryl bromides[4] (eq. 4)
  • Catalyst for Buchwald-Hartwig amination reaction of aryl halides[4] (eq. 5)
  • Catalyst for Negishi coupling reaction[5] (eq. 6)
Catalyst for C-C, C-N, and N-S bond formation.

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Pyr. Sol. 1 - Skin Corr. 1B - Water-react 2

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric A Bercot et al.
Organic letters, 10(22), 5251-5254 (2008-10-29)
A diastereoselective palladium-catalyzed arylation of 4-substituted cyclohexyl esters has been developed. The reaction proceeds at room temperature in the presence of [(t-Bu3P)PdBr]2 providing products in up to 37:1 dr.
Palladium-catalyzed gamma-arylation of alpha,beta-unsaturated esters from silyl ketene acetals.
David S Huang et al.
Angewandte Chemie (International ed. in English), 49(33), 5757-5761 (2010-07-08)
Chad C Eichman et al.
The Journal of organic chemistry, 74(10), 4005-4008 (2009-04-07)
A catalytic amount of zinc chloride in combination with a palladium catalyst ligated by a monodentate phosphine allows the coupling of aryl and alkyl thiols with aryl bromides in high yields. The addition of zinc chloride to a palladium catalyst
Takuo Hama et al.
Journal of the American Chemical Society, 128(15), 4976-4985 (2006-04-13)
The intermolecular alpha-arylation and vinylation of amides by palladium-catalyzed coupling of aryl bromides and vinyl bromides with zinc enolates of amides is reported. Reactions of three different types of zinc enolates have been developed. The reactions of aryl halides occur
Unparalleled rates for the activation of aryl chlorides and bromides: coupling with amines and boronic acids in minutes at room temperature.
James P Stambuli et al.
Angewandte Chemie (International ed. in English), 41(24), 4746-4748 (2002-12-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service