Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

673560

Sigma-Aldrich

4-(6-Mercaptohexyloxy)benzyl alcohol

Synonym(s):

4-Hydroxymethyl-1-(6-mercaptohexyloxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O2S
CAS Number:
Molecular Weight:
240.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

44-49 °C

storage temp.

−20°C

SMILES string

OCc1ccc(OCCCCCCS)cc1

InChI

1S/C13H20O2S/c14-11-12-5-7-13(8-6-12)15-9-3-1-2-4-10-16/h5-8,14,16H,1-4,9-11H2

InChI key

HHHRERFRNRFXOV-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service